Anti-SARS-CoV-2 Spike RBD Antibody, Chimeric mAb, Human IgG1 S1N-M122 is a chimeric monoclonal antibody combining the constant domai ns of the human IgG1 molecule with mouse variable regions. The variable region was obtained from a mouse immunized wi 1 * 100 µG
Catalog # ACRBS1N-M122-100UG
Supplier: ACRO Biosystems
Frequently Bought Together