FITC-Labeled Human CD8ACD8B Heterodimer Protein, His TagTag Free CDA-HF2W4 is expressed from human 293 cells HEK293. It contains AA Ser 22 - Asp 182 CD8A Leu 22 - Pro 170 CD8B Accession P01732-1 CD8A P10966-1 CD8B. It is the FITC labeled form of Het 1 * 200 µG
Catalog # ACRBCDA-HF2W4200UG
Supplier: ACRO Biosystems
Frequently Bought Together