ContactUsLinkComponent
Biotinylated Human CD8 alpha, His,Avitag CDA-H82E3 is expressed from human 293 cells HEK293. It contains AA Ser 22 - Asp 182 Accessi on P01732-1. 1 * 200 µG
Catalog # ACRBCDA-H82E3200UG
Supplier:  ACRO Biosystems
Biotinylated Human CD8 alpha, His,Avitag CDA-H82E3 is expressed from human 293 cells HEK293. It contains AA Ser 22 - Asp 182 Accessi on P01732-1. 1 * 200 µG
Catalog # ACRBCDA-H82E3200UG
Supplier:  ACRO Biosystems
Frequently Bought Together