ContactUsLinkComponent
PE-Labeled Human CD19 20-291, His Tag CD9-HP2H3 is produced via conjugation of PE to Human CD19 20-291, His Tag under optimal condit ions. Human CD19 20-291, His Tag is expressed in HEK293 cells containing AA Pro 20 - Lys 291 Accession P15391-1. 1 * 200 Tests
Catalog # ACRBCD9HP2H3200TST
Supplier:  ACROBIOSYSTEMS
PE-Labeled Human CD19 20-291, His Tag CD9-HP2H3 is produced via conjugation of PE to Human CD19 20-291, His Tag under optimal condit ions. Human CD19 20-291, His Tag is expressed in HEK293 cells containing AA Pro 20 - Lys 291 Accession P15391-1. 1 * 200 Tests
Catalog # ACRBCD9HP2H3200TST
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together