ContactUsLinkComponent
The biotinylated ACE2 protein was conjugated to streptavidin magnetic beads. This pre-coupled magnetic bead product can capture the SARS-CoV-2 S1 proteinSARS-CoV-2 S protein RBD from various assay systems. The beads are in uniform size, narrow size d 1 * 2 mg
Catalog # ACRBMBS-K013-2MG
Supplier:  ACROBIOSYSTEMS
The biotinylated ACE2 protein was conjugated to streptavidin magnetic beads. This pre-coupled magnetic bead product can capture the SARS-CoV-2 S1 proteinSARS-CoV-2 S protein RBD from various assay systems. The beads are in uniform size, narrow size d 1 * 2 mg
Catalog # ACRBMBS-K013-2MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together