ContactUsLinkComponent
Biotinylated SARS-CoV-2 S1 protein D614G, His,Avitag™ S1N-C82E3 is expressed from human 293 cells HEK293. It contains AA Val 16 - Ar g 685 Accession QHD43416.1 D614G. 1 * 25 µG
Catalog # ACRBS1N-C82E3-25UG
Supplier:  ACRO Biosystems
Biotinylated SARS-CoV-2 S1 protein D614G, His,Avitag™ S1N-C82E3 is expressed from human 293 cells HEK293. It contains AA Val 16 - Ar g 685 Accession QHD43416.1 D614G. 1 * 25 µG
Catalog # ACRBS1N-C82E3-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together