ContactUsLinkComponent
Biotinylated SARS-CoV-2 S protein, His, Avitag, Super stable trimer SPN-C82E9 is the ectodomain of SARS-CoV-2 S protein which contai ns AA Val 16 - Pro 1213 Accession QHD43416.1. The recombinant protein is expressed from human 293 cells HEK293 with m 1 * 200 µG
Catalog # ACRBSPN-C82E9200UG
Supplier:  ACROBIOSYSTEMS
Biotinylated SARS-CoV-2 S protein, His, Avitag, Super stable trimer SPN-C82E9 is the ectodomain of SARS-CoV-2 S protein which contai ns AA Val 16 - Pro 1213 Accession QHD43416.1. The recombinant protein is expressed from human 293 cells HEK293 with m 1 * 200 µG
Catalog # ACRBSPN-C82E9200UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together