ContactUsLinkComponent
Human BAFFR Protein, His Tag BAR-H52H3 is expressed from human 293 cells HEK293. It contains AA Ser 7 - Ala 71 Accession Q96RJ3-1. 1 * 100 µG
Catalog # ACRBBAR-H52H3100UG
Supplier:  ACROBIOSYSTEMS
Human BAFFR Protein, His Tag BAR-H52H3 is expressed from human 293 cells HEK293. It contains AA Ser 7 - Ala 71 Accession Q96RJ3-1. 1 * 100 µG
Catalog # ACRBBAR-H52H3100UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together