ContactUsLinkComponent
Biotinylated Human PVRIG, Fc,Avitag™ HPLC verified PVG-H82F9 is expressed from human 293 cells HEK293. It contains AA Thr 41 - Asp 1 71 Accession Q6DKI7-1. 1 * 200 µG
Catalog # ACRBPVG-H82F9200UG
Supplier:  ACRO Biosystems
Biotinylated Human PVRIG, Fc,Avitag™ HPLC verified PVG-H82F9 is expressed from human 293 cells HEK293. It contains AA Thr 41 - Asp 1 71 Accession Q6DKI7-1. 1 * 200 µG
Catalog # ACRBPVG-H82F9200UG
Supplier:  ACRO Biosystems
Frequently Bought Together