The pre-coupled magnetic beads coupled with biotinylated SARS-CoV-2 Spike trimerD614G protein to streptavidin conjugated magnetic be ads, which can capture the Anti- SARS-CoV-2 antibody or ACE2 protein from cell or serum sample. The beads are in unif 1 * 10 mg
Catalog # ACRBMBS-K016-10MG
Supplier: ACROBIOSYSTEMS
Frequently Bought Together