ContactUsLinkComponent
HCoV-NL63 Nucleocapsid, His Tag NUN-V5146 is expressed from E.coli cells. It contains AA Met 1 - His 377 Accession Q6Q1R8-1. 1 * 1 mg
Catalog # ACRBNUN-V5146-1MG
Supplier:  ACRO Biosystems
HCoV-NL63 Nucleocapsid, His Tag NUN-V5146 is expressed from E.coli cells. It contains AA Met 1 - His 377 Accession Q6Q1R8-1. 1 * 1 mg
Catalog # ACRBNUN-V5146-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together