PE-Labeled Monoclonal Anti-FMC63 scFv Antibody, Mouse IgG1 Cat. No. FM3-HPY53 is produced via conjugation of PE to Monoclonal Anti-F MC63 scFv Antibody, Mouse IgG1 under optimal conditions. 1 * 200 Tests
Catalog # ACRBFM3HPY53200TST
Supplier: ACRO Biosystems
Frequently Bought Together