Auftragserfassung
Switzerland
ContactUsLinkComponent
CTLA-4 CD152 HUMAN HIS BLI VERIFIED 1 * 100 µG
Catalog # ACRBCT4-H52H9100UG
Supplier:  ACRO Biosystems
CTLA-4 CD152 HUMAN HIS BLI VERIFIED 1 * 100 µG
Catalog # ACRBCT4-H52H9100UG
Supplier:  ACRO Biosystems
Frequently Bought Together