ContactUsLinkComponent
Biotinylated SARS-CoV-2 S2 protein, His,Avitag™ S2N-C52E8 is expressed from human 293 cells HEK293. It contains AA Ser 686 - Pro 121 3 Accession QHD43416.1. 1 * 200 µG
  ACRBS2N-C52E8200UG
Biotinylated SARS-CoV-2 S2 protein, His,Avitag™ S2N-C52E8 is expressed from human 293 cells HEK293. It contains AA Ser 686 - Pro 121 3 Accession QHD43416.1. 1 * 200 µG
  ACRBS2N-C52E8200UG
Frequently Bought Together