ContactUsLinkComponent
FITC-Labeled Human CEACAM-5, His Tag CE5-HF254 is expressed from human 293 cells HEK293. It contains AA Lys 35 - Ala 685 Accession P06731-1. It is the FITC labeled form of Human CEACAM-5, His Tag Cat. No.CE5-H5226. 1 * 25 µG
Catalog # ACRBCE5-HF254-25UG
Supplier:  ACROBIOSYSTEMS
FITC-Labeled Human CEACAM-5, His Tag CE5-HF254 is expressed from human 293 cells HEK293. It contains AA Lys 35 - Ala 685 Accession P06731-1. It is the FITC labeled form of Human CEACAM-5, His Tag Cat. No.CE5-H5226. 1 * 25 µG
Catalog # ACRBCE5-HF254-25UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together