ContactUsLinkComponent
FITC-Labeled Human VEGF R2, His Tag VE2-HF254 is expressed from human 293 cells HEK293. It contains AA Ala 20 - Glu 764 Accession A AI31823.1. It is the FITC labeled form of Human VEGF R2, His Tag Cat. No.KDR-H5227. 1 * 25 µG
Catalog # ACRBVE2-HF254-25UG
Supplier:  ACROBIOSYSTEMS
FITC-Labeled Human VEGF R2, His Tag VE2-HF254 is expressed from human 293 cells HEK293. It contains AA Ala 20 - Glu 764 Accession A AI31823.1. It is the FITC labeled form of Human VEGF R2, His Tag Cat. No.KDR-H5227. 1 * 25 µG
Catalog # ACRBVE2-HF254-25UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together