ContactUsLinkComponent
FITC-Labeled Human B7-H3 4Ig, His Tag B7B-HF2E7 is expressed from human 293 cells HEK293. It contains AA Gly 27 - Thr 461 Accession Q5ZPR3-1. It is the FITC labeled form of Human B7-H3 4Ig, His Tag Cat. No.B7B-H52E7. 1 * 200 µG
Catalog # ACRBB7B-HF2E7200UG
Supplier:  ACROBIOSYSTEMS
FITC-Labeled Human B7-H3 4Ig, His Tag B7B-HF2E7 is expressed from human 293 cells HEK293. It contains AA Gly 27 - Thr 461 Accession Q5ZPR3-1. It is the FITC labeled form of Human B7-H3 4Ig, His Tag Cat. No.B7B-H52E7. 1 * 200 µG
Catalog # ACRBB7B-HF2E7200UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together