ContactUsLinkComponent
Biotinylated Human CD7, His,Avitag CD7-H82E4 is expressed from human 293 cells HEK293. It contains AA Ala 26 - Pro 180 Accession P0 9564-1. 1 * 200 µG
Catalog # ACRBCD7-H82E4200UG
Supplier:  ACRO Biosystems
Biotinylated Human CD7, His,Avitag CD7-H82E4 is expressed from human 293 cells HEK293. It contains AA Ala 26 - Pro 180 Accession P0 9564-1. 1 * 200 µG
Catalog # ACRBCD7-H82E4200UG
Supplier:  ACRO Biosystems
Frequently Bought Together