ContactUsLinkComponent
Human IL-34, His Tag IL4-H52H3 is expressed from human 293 cells HEK293. It contains AA Asn 21 - Pro 242 Accession Q6ZMJ4-1. 1 * 100 µG
Catalog # ACRBIL4-H52H3100UG
Supplier:  ACRO Biosystems
Human IL-34, His Tag IL4-H52H3 is expressed from human 293 cells HEK293. It contains AA Asn 21 - Pro 242 Accession Q6ZMJ4-1. 1 * 100 µG
Catalog # ACRBIL4-H52H3100UG
Supplier:  ACRO Biosystems
Frequently Bought Together