ContactUsLinkComponent
Biotinylated Human Alpha-Synuclein, His,Avitag ALN-H82H8 is expressed from E.coli cells. It contains AA Met 1 - Ala 140 Accession P 37840-1. 1 * 200 µG
Catalog # ACRBALN-H82H8200UG
Supplier:  ACRO Biosystems
Biotinylated Human Alpha-Synuclein, His,Avitag ALN-H82H8 is expressed from E.coli cells. It contains AA Met 1 - Ala 140 Accession P 37840-1. 1 * 200 µG
Catalog # ACRBALN-H82H8200UG
Supplier:  ACRO Biosystems
Frequently Bought Together