ContactUsLinkComponent
Human HBEGF, His Tag HBF-H52H3 is expressed from human 293 cells HEK293. It contains AA Asp 63 - Leu 148 Accession Q99075-1. 1 * 100 µG
Catalog # ACRBHBF-H52H3100UG
Anbieter:  ACRO Biosystems
Human HBEGF, His Tag HBF-H52H3 is expressed from human 293 cells HEK293. It contains AA Asp 63 - Leu 148 Accession Q99075-1. 1 * 100 µG
Catalog # ACRBHBF-H52H3100UG
Anbieter:  ACRO Biosystems
Frequently Bought Together