ContactUsLinkComponent
SARS-CoV-2 S1 protein L18F, D80A, D215G, R246I, K417N, E484K, N501Y, D614G, His Tag S1N-C52Hm is expressed from human 293 cells HEK2 93. It contains AA Val 16 - Arg 685 L18F, D80A, D215G, R246I, K417N, E484K, N501Y, D614G Accession QHD43416.1 L18F, 2 1 * 100 µG
Catalog # ACRBS1N-C52HM100UG
Supplier:  ACROBIOSYSTEMS
SARS-CoV-2 S1 protein L18F, D80A, D215G, R246I, K417N, E484K, N501Y, D614G, His Tag S1N-C52Hm is expressed from human 293 cells HEK2 93. It contains AA Val 16 - Arg 685 L18F, D80A, D215G, R246I, K417N, E484K, N501Y, D614G Accession QHD43416.1 L18F, 2 1 * 100 µG
Catalog # ACRBS1N-C52HM100UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together