ContactUsLinkComponent
PiColorLock phosphate detection reagent changes colour in the presence of inorganic phosphate (Pi). This can be exploited to measure any enzyme that generates Pi including ATPases, GTPases, phosphatases, heat-shock proteins and DNA unwinding proteins 1 * 1 KIT
Catalog # NOVU303-0125
Supplier:  Novus Biologicals
PiColorLock phosphate detection reagent changes colour in the presence of inorganic phosphate (Pi). This can be exploited to measure any enzyme that generates Pi including ATPases, GTPases, phosphatases, heat-shock proteins and DNA unwinding proteins 1 * 1 KIT
Catalog # NOVU303-0125
Supplier:  Novus Biologicals
Frequently Bought Together