Electro-valve block and connector 1 * 1 items
Catalog # HACHLZE165
Supplier: Hach
Frequently Bought Together
Accessories for SP 100
Supplier: Socorex Isba
ThermoMixer® C Thermomixer
Supplier: EPPENDORF
Digital pocket refractometers, PAL series
Supplier: Atago
Accessories for OxiTop® and OxiTop® Control
Supplier: WTW
pH/mV/°C meters, handheld, pH 3110 / 3310
Supplier: WTW
Accessories for Mettler balances
Supplier: Mettler - Toledo
VWR®, Square Cuvettes
Supplier: VWR Collection
Accessories for spiral plater/diluter, easySpiral®
Supplier: INTERSCIENCE
VWR®, Single-Use Heads for Air Samplers
Supplier: VWR Collection
Multi-parameter meters, handheld, 3320
Supplier: WTW
VWR® pHenomenal® MU 6100 H Multi-Parameter Meter (pH/Conductivity/DO), Handheld
Supplier: VWR Collection
ESCHERICHIA COLI WDCM 00013 VITROIDS CRM 1 * 10 items
Supplier: Merck
Components for spectrophotometric colorimeters, PFX<i>i</i>-195/880/950/995 series, Lovibond®
Supplier: Lovibond Tintometer
Gas detection tubes for carbon dioxide
Supplier: Dräger
SmartBlocks for ThermoMixer® C and ThermoStat C
Supplier: EPPENDORF
Accessories for titrators
Supplier: Mettler - Toledo