ContactUsLinkComponent
Glove R085 R-Flex seamless breathable HPPE knit palm-dip with sandy foam finish featuring fleece liner, cut protection and high dexterity. Touch Screen compatible on first two fingers and thumb. Size 8 1 * 8 PAIR
Catalog # ANSER085080
Supplier:  Ansell
Glove R085 R-Flex seamless breathable HPPE knit palm-dip with sandy foam finish featuring fleece liner, cut protection and high dexterity. Touch Screen compatible on first two fingers and thumb. Size 8 1 * 8 PAIR
Catalog # ANSER085080
Supplier:  Ansell
Frequently Bought Together