ContactUsLinkComponent
A control for BSA-fatty acid complexes has been used as a control for experiments using BSA-fatty acid complexes to deliver fatty acids to cells in culture 1 * 5 mL
Catalog # CAYM29556-5
Supplier:  Cayman Chemical
A control for BSA-fatty acid complexes has been used as a control for experiments using BSA-fatty acid complexes to deliver fatty acids to cells in culture 1 * 5 mL
Catalog # CAYM29556-5
Supplier:  Cayman Chemical
Frequently Bought Together