ContactUsLinkComponent
COD100.L1,,Chemical Oxygen Demand COD 100 mg/l Calibration Standard. Duree de conservation : 24 mois. Store in a refrigerator at temperatures between 2C to 8C,,100 ml bottle,, 1 * 100 mL
Catalog # ACSDCOD100.L1
Supplier:  CPACHEM
COD100.L1,,Chemical Oxygen Demand COD 100 mg/l Calibration Standard. Duree de conservation : 24 mois. Store in a refrigerator at temperatures between 2C to 8C,,100 ml bottle,, 1 * 100 mL
Catalog # ACSDCOD100.L1
Supplier:  CPACHEM
Frequently Bought Together