Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Glutamine

Glutamine

L-glutamic acid (powder), 1 kg, 1 kg
Catalog # P1011-1KG
Supplier:   Biowest
Glutamine
Catalog # P1011-1KG
Supplier:  Biowest
CAS Number:  
Glutamine
Glutamine

L-glutamic acid (powder), 1 kg, 1 kg

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 kg
Product Details & Documents

L-glutamine is an essential amino acid required by virtually all mammalian and insect cells grown in culture. It is a crucial component of many cell culture media and serves as a major energy source for cells in culture. L-glutamine is very stable as a dry powder and as a frozen solution. However, in liquid media or stock solutions, L-glutamine can degrade relatively rapidly. L-glutamine is also more labile in cell culture solution than other amino acids.

Dipeptide derivatives of L-glutamine (stable glutamine) prevent the intramolecular cyclisation reaction associated with solutions of L-glutamine. These derivatives are therefore stable in solution and allow the formulation of cell culture media containing L-glutamine that may be stored at 4 °C for extended periods. Solutions containing these derivatives can even be autoclaved without appreciable degradation of the product (30 minutes at 121 °C results in <5% loss of the product).

The dipeptide derivatives are metabolised within the cells to yield L-glutamine plus the second amino acid. This results in more consistent delivery of L-glutamine to cells and avoids toxic build-up of ammonia in cell cultures. This feature can be especially important for ammonia-sensitive cell lines.

Documents

Specifications

Specifications

  • Pk:
    1 kg
  • Description:
    L-glutamic acid (powder)
  • Size:
    1 kg

Specifications