Order Entry
Switzerland
Orders LinkContactUsLinkComponent
AccuScript high-fidelity reverse transcriptase

AccuScript high-fidelity reverse transcriptase

High fidelity reverse transcriptase for long target lengths. Ideal for low RNA input and high specificity. Shorter PCR extension times with PfuUltra II with high accuracy. Kit includes dNTP, dTT, oligodT and random primers. Capable of amplifying cDNA of 0,1 - 9,6 kb in length ;10 - 1000 ng of total RNA or 0,1 - 10 ng of poly(A)+ RNA input, 50 reactions
Catalog # AGLS600180
Supplier:   AGILENT
AccuScript high-fidelity reverse transcriptase
Catalog # AGLS600180
Supplier:  AGILENT
CAS Number:  
AccuScript high-fidelity reverse transcriptase

High fidelity reverse transcriptase for long target lengths. Ideal for low RNA input and high specificity. Shorter PCR extension times with PfuUltra II with high accuracy. Kit includes dNTP, dTT, oligodT and random primers. Capable of amplifying cDNA of 0,1 - 9,6 kb in length ;10 - 1000 ng of total RNA or 0,1 - 10 ng of poly(A)+ RNA input, 50 reactions

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

AccuScript High-Fidelity Reverse Transcriptase delivers the highest RT accuracy currently available.

  • MMLV-RT derivative combined with 3 to 5’ exonuclease reduces errors
  • High yields of full length cDNA up to 20 kb;RT-PCR kit synthesises cDNA from 10 to 1000 ng of total RNA or 0,1 to 10 ng of poly(A)+ RNA
  • Stringent quality control procedures ensure high yields of premium cDNA

AccuScript High-Fidelity Reverse Transcriptase (RT) delivers 3,7 times more accuracy than other commercially available RTs and 6,6 times more accuracy than AMV RT. The AccuScript PfuUltra II RT-PCR kit provides a complete system in applications that demand high fidelity, specificity, and throughput.

Caution: For research use only. Not for use in diagnostic procedures.

Specifications

Specifications

  • Details:
    High fidelity reverse transcriptase for long target lengths. Ideal for low RNA input and high specificity. Shorter PCR extension times with PfuUltra II with high accuracy. Kit includes dNTP, dTT, oligodT and random primers. Capable of amplifying cDNA of 0,1 - 9,6 kb in length ;10 - 1000 ng of total RNA or 0,1 - 10 ng of poly(A)+ RNA input
  • No. of reactions:
    50 reactions

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.