Precellys® Evolution Touch Homogeniser
Combo Precellys Evolution and Cryolys
432-0454
The Precellys Evolution Touch is both flexible and efficient, giving users the ability to grind, homogenise or lyse any kind of sample.
- Use six different tube sizes (from 0,3 to 50 ml) and prepare between 1 and 96 tubes at the same time
- From 4500 to 10000 rpm (increments of 100 rpm)
- Automatic tube locking system
- The software allows users to save an unlimited number of protocols
- Run up to 10 cycles
Easy to use with its new intuitive touchscreen interface, designed to be used with gloves, researchers can easily set up the Precellys Evolution Touch. The Precellys Evolution Touch puts the power of homogenisation into the hands of researchers and scientists.
Tube holders and other accessories for the Precellys Evolution are fully compatible with the Precellys Evolution Touch.
: E and CFR 47 part 15 certified.
- Description:Combo Precellys Evolution and Cryolys
- Seq:02
Accessories
- DescriptionFor
- BERTS002275PEVO0A0Metal tube holder pack (1×metal tube blocking plate and 1×metal tube holder)Precellys® Evolution1 * 1 ST
- DescriptionFor
- 432-0306Empty metal tubes set for Precellys® Evolution homogenisers, 6×7 ml metal tubes, tube holder and blocking platePersonalised filling1 * 1 ST - 3 340.00 SFr.
- DescriptionFor
- 432-0346Cryolys® Evolution, ONLY works with dry ice and it is placed on top of the Precellys Evolution.Cooling samples while using Precellys® Evolution homogeniser1 * 1 ST - 7 980.00 SFr.
- DescriptionFor
- BERTS002474PEVO0A050 ml holder pack (1×50 ml tube holder working as blocking plate)Precellys® Evolution1 * 1 ST - 1 410.00 SFr.



