ContactUsLinkComponent
12 components 100mg/l each of Amidosulfuron [CAS:120923-37-7] Bensulfuron-methyl [CAS:83055-99-6] Cinosulfuron [CAS:94593-91-6] Chlorsulfuron [CAS:64902-72-3] Flazasulfuron [CAS:104040-78-0] Iodosulfuron-methyl-sodium [CAS:144550-36-7] [ 1 * 5 Ampoul
  CMPLRD0013071.5
12 components 100mg/l each of Amidosulfuron [CAS:120923-37-7] Bensulfuron-methyl [CAS:83055-99-6] Cinosulfuron [CAS:94593-91-6] Chlorsulfuron [CAS:64902-72-3] Flazasulfuron [CAS:104040-78-0] Iodosulfuron-methyl-sodium [CAS:144550-36-7] [ 1 * 5 Ampoul
  CMPLRD0013071.5
Frequently Bought Together