Tip robotic 250l Beckman style Low Retention rack sterile (960tips/pack, 5 packs/case) 1 * 4.800 items
Catalog # GEBSAT250B-9-NS-LB
Supplier: SHANGHAI GENE ERA BIO SCIENCE
Tip robotic 250l Beckman style Low Retention rack sterile (960tips/pack, 5 packs/case) 1 * 4.800 items
Catalog # GEBSAT250B-9-NS-LB
Supplier: SHANGHAI GENE ERA BIO SCIENCE
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR® Facial Tissues
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection