ContactUsLinkComponent
HAAKE Viscotester 3 R Viscometer Base viscometer in carrying case with instrument holder for lab stand, one measuring cup (No. 3), three rotors (No. 1, 2 and 3), four rechargeable batteries and one power supply adapter Viscosity range : R Version (Fo 1 * 1 items
Katalognr. HAAK399-0112
Anbieter:  HAAKE
HAAKE Viscotester 3 R Viscometer Base viscometer in carrying case with instrument holder for lab stand, one measuring cup (No. 3), three rotors (No. 1, 2 and 3), four rechargeable batteries and one power supply adapter Viscosity range : R Version (Fo 1 * 1 items
Katalognr. HAAK399-0112
Anbieter:  HAAKE
Frequently Bought Together