ContactUsLinkComponent
CHROMABOND columns HR-XAW weak mixed-mode polymer-based anion exchanger volume: 3 mL, content of sorbent: 60 mg material: PP, with PE filter elements packed in 5 bags with 50 pcs. pack of 250 1 * 250 items
Catalog # MANA730747.250
Supplier:  MACHEREY-NAGEL
CHROMABOND columns HR-XAW weak mixed-mode polymer-based anion exchanger volume: 3 mL, content of sorbent: 60 mg material: PP, with PE filter elements packed in 5 bags with 50 pcs. pack of 250 1 * 250 items
Catalog # MANA730747.250
Supplier:  MACHEREY-NAGEL
Frequently Bought Together