di-Sodium hydrogen phosphate anhydrous EMPROVE ESSENTIAL Ph Eur,BP,USP,JPE,E 339 1 * 12 kg
Catalog # 1.06585.9013
Supplier: MERCK PRODUCTION CHEMICALS
di-Sodium hydrogen phosphate anhydrous EMPROVE ESSENTIAL Ph Eur,BP,USP,JPE,E 339 1 * 12 kg
Catalog # 1.06585.9013
Supplier: MERCK PRODUCTION CHEMICALS
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Potassium dihydrogen phosphate, crystallised, EMPROVE® ESSENTIAL Ph. Eur., BP, JPC, NF, E340, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals