1- (2-hydroxyethyl)-5- methyl-1,2,3,4- tetrahydropyrimidi ne-2,4-dione, CAS 22441-51-6, MFCD21106605, 95% 1 * 1 g
Catalog # UKROBBV-39900110
Supplier: UKRORGSYNTHESIS
1- (2-hydroxyethyl)-5- methyl-1,2,3,4- tetrahydropyrimidi ne-2,4-dione, CAS 22441-51-6, MFCD21106605, 95% 1 * 1 g
Catalog # UKROBBV-39900110
Supplier: UKRORGSYNTHESIS
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
VWR® Facial Tissues
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection