Vanadium(V) oxide ≥98%, GPR RECTAPUR®
Plastic bottle, 500 g
Supplier: VWR Chemicals
Vanadium(V) oxide ≥98%, GPR RECTAPUR®
Catalog # 87054.260
Supplier: VWR Chemicals
CAS Number: 1314-62-1
Jump to:
Product Details & Documents
Synonyms: Vanadium pentoxide, di-Vanadium pentaoxide
- CAS Number:1314-62-1
- Formula:O₅V₂
- MDL Number:MFCD00011457
- Molecular Weight:181.88 g/mol
- UN:2862
- ADR:6.1,III
Documents
Certificates
Search by Lot Numberspec_VWRC87054.180 spec_VWRC87054.260 spec_VWRC87054.260
Product specification
Specifications
Specification Test Results
- Assay:≥ 98 %
- Appearance:Brown crystalline powder
- Total N (Nitrogen):≤ 0.1 %
- Cl (Chloride):≤ 0.1 %
- PO₄ (Phosphate):≤ 0.1 %
- SO₄ (Sulphate):≤ 0.1 %
- Fe (Iron):≤ 0.1 %
Specifications
- Pack type:Plastic bottle
- Pk:500 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



