ContactUsLinkComponent
Converter Piezoelectric lead zirconate titanate crystals (PZT) Part No. CV 188 with pulsing button. Diameter: 32 mm Length: 146 mm, Weight: 3 340 g, Cable length CV 188: 1.8 m 1 * 1 items
Catalog # SONICV188
Supplier:  SONICS MATERIALS
Converter Piezoelectric lead zirconate titanate crystals (PZT) Part No. CV 188 with pulsing button. Diameter: 32 mm Length: 146 mm, Weight: 3 340 g, Cable length CV 188: 1.8 m 1 * 1 items
Catalog # SONICV188
Supplier:  SONICS MATERIALS
Frequently Bought Together