Micro electrode SCPpHT-MIC-AMF-3M-DIN-N temp. sensor Pt 1000, 3 m cable with DIN and banana plug, glass shaft, Pt junction,electr. KCl 3 mol/l, Silamid-reference system with silver ion barrier and double-electrolyte, cylindrical membrane, A glass, le 1 * 1 items
Catalog # SCOI285101335
Supplier: SI Analytics
Frequently Bought Together