ContactUsLinkComponent
Protecta Shock Absorbing Lanyard 140 kg capacity, Rope Single Leg, adjustable length 1,5 - 2,0 m, screw gate carabiner, 17 mm openin g, 25 kN breaking strength, screw gate carabiner, 17 mm opening, 25 kN breaking strength 1 * 1 items
Catalog # CPSD1260334
Supplier:  CAPITAL SAFETY
Protecta Shock Absorbing Lanyard 140 kg capacity, Rope Single Leg, adjustable length 1,5 - 2,0 m, screw gate carabiner, 17 mm openin g, 25 kN breaking strength, screw gate carabiner, 17 mm opening, 25 kN breaking strength 1 * 1 items
Catalog # CPSD1260334
Supplier:  CAPITAL SAFETY
Frequently Bought Together