ContactUsLinkComponent
Protecta Shock Absorbing Lanyard 140 kg capacity, Elastic Single Leg, self-locking twist lock carabiner, 18 mm opening, 25 kN breaki ng strength, self-locking scaffold hook, 50 mm opening, 23 kN breaking strength 1 * 1 items
Catalog # CPSD1260328
Supplier:  CAPITAL SAFETY
Protecta Shock Absorbing Lanyard 140 kg capacity, Elastic Single Leg, self-locking twist lock carabiner, 18 mm opening, 25 kN breaki ng strength, self-locking scaffold hook, 50 mm opening, 23 kN breaking strength 1 * 1 items
Catalog # CPSD1260328
Supplier:  CAPITAL SAFETY
Frequently Bought Together