ContactUsLinkComponent
Bottle reagent, capacity 1l, color brown, narrow mouth, ground in flat stopper, diameter 106.5mm, height 199mm, SJ 29/32. 1 * 10 items
Catalog # KAVAN632414202940
Supplier:  Kavalier
Bottle reagent, capacity 1l, color brown, narrow mouth, ground in flat stopper, diameter 106.5mm, height 199mm, SJ 29/32. 1 * 10 items
Catalog # KAVAN632414202940
Supplier:  Kavalier
Frequently Bought Together