Silver AAS standards, (CRMs), Fluka™
Silver, Ag, 10000 mg/L, 20% HNO₃, Plastic bottle, 100 mL
Catalog # HONCE4100-100ML
Supplier: Honeywell Chemicals
Silver AAS standards, (CRMs), Fluka™
Catalog # HONCE4100-100ML
Supplier: Honeywell Chemicals
CAS Number:
Jump to:
Product Details & Documents
Certified Reference Materials (CRMs)
- Traceable to SRM from NIST
: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).
Specifications
Specifications
- Concentration:10000 mg/L
- Matrix:20% HNO₃
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Silver
- Symbol:Ag
- Cat. No.:HONCE4100-100ML



