Rubidium AAS Standards, (CRMs), Fluka™
Rubidium, Rb, 1000 mg/L, 2% HNO₃, Plastic bottle, 100 mL
HONCE4059-100ML
Certified Reference Materials (CRMs)
- Traceable to SRM from NIST
: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).
- Concentration:1000 mg/L
- Matrix:2% HNO₃
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Rubidium
- Symbol:Rb



