Silver AAS standards, (CRMs), Fluka™
Silver, Ag, 1000 mg/L, 2% HNO₃, Plastic bottle, 500 mL
Catalog # HONCE4000-500ML
Supplier: Honeywell Chemicals
Silver AAS standards, (CRMs), Fluka™
Catalog # HONCE4000-500ML
Supplier: Honeywell Chemicals
CAS Number:
Jump to:
Product Details & Documents
Certified Reference Materials (CRMs)
- Traceable to SRM from NIST
Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).
Specifications
Specifications
- Concentration:1000 mg/L
- Matrix:2% HNO₃
- Pack type:Plastic bottle
- Pk:500 mL
- Element:Silver
- Symbol:Ag



