ContactUsLinkComponent
YMC Triart Bio C4 Capillary Column,30 nm,s-3um,150*0.3mm ID 1/3
  YMCATB30S03-15H0RU
 :  YMC
YMC Triart Bio C4 Capillary Column,30 nm,s-3um,150*0.3mm ID 1/3
  YMCATB30S03-15H0RU
 :  YMC
Frequently Bought Together