ContactUsLinkComponent
HighPurS-CERT Sodium Nitrite (NaNO2)- min. 99.99+% - ISO 17034 and ISO/IEC 17025 certified. Duree de conservation : 36 mois. Store under normal laboratory conditions, at temperatures between 15 to 25C 1 * 100 g
Katalognr. ACSDPSNANIC1
Anbieter:  CPACHEM
HighPurS-CERT Sodium Nitrite (NaNO2)- min. 99.99+% - ISO 17034 and ISO/IEC 17025 certified. Duree de conservation : 36 mois. Store under normal laboratory conditions, at temperatures between 15 to 25C 1 * 100 g
Katalognr. ACSDPSNANIC1
Anbieter:  CPACHEM
Zugehörige Informationen
Frequently Bought Together