Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Creatinine NF

Creatinine NF

Poly pail, 1 kg
Catalog # SPCMC1403-1KGBL
Supplier:   Spectrum Chemical
Creatinine NF
Catalog # SPCMC1403-1KGBL
Supplier:  Spectrum Chemical
CAS Number:  60-27-5
Creatinine NF

Poly pail, 1 kg

CAS Number:  60-27-5
Price
SFr. 1’150.00
Log in for availability
1 * 1 kg
Product Details & Documents

All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Synonyms:  2-Amino-1-methyl-1,5-dihydro-4H-imidazol-4-one
  • CAS Number:60-27-5
  • Formula:C₄H₇N₃O
  • MDL Number:MFCD00059730
  • Molecular Weight:113.12 g/mol
  • Storage Temperature:Ambient
  • EINECS:200-466-7
Specifications

Specification Test Results

  • Assay:
    98.5% Min.

Specifications

  • Pack type:
    Poly pail
  • Pk:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.