Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Creatinine NF

Poly pail, 12 kg
Catalog # SPCMC1403-12KGBL
Supplier:   Spectrum Chemical
Creatinine NF
Catalog # SPCMC1403-12KGBL
Supplier:  Spectrum Chemical
CAS Number:  60-27-5
Creatinine NF

Poly pail, 12 kg

CAS Number:  60-27-5
Price
SFr. 4’280.00
Log in for availability
1 * 12 kg
Product Details & Documents

All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Synonyms:  2-Amino-1-methyl-1,5-dihydro-4H-imidazol-4-one
  • CAS Number:60-27-5
  • Formula:C₄H₇N₃O
  • MDL Number:MFCD00059730
  • Molecular Weight:113.12 g/mol
  • Storage Temperature:Ambient
  • EINECS:200-466-7
Specifications

Specification Test Results

  • Assay:
    98.5% Min.

Specifications

  • Pack type:
    Poly pail
  • Pk:
    12 kg

Specifications