ContactUsLinkComponent
Dimethyl ether [CAS:115-10-6] 4500mg/l Formic acid methyl ester [CAS:107-31-3] 7000mg/l Diethylether [CAS:60-29-7] 4500mg/l Is opropyl ether [CAS:108-20-3] 4500mg/l Tetrahydrofuran [CAS:109-99-9] 3000mg/l Dioxan [CAS:123-91-1] 600mg/l Dib 1 * 10 mL
Catalog # CMPLRD04468610.2
Dimethyl ether [CAS:115-10-6] 4500mg/l Formic acid methyl ester [CAS:107-31-3] 7000mg/l Diethylether [CAS:60-29-7] 4500mg/l Is opropyl ether [CAS:108-20-3] 4500mg/l Tetrahydrofuran [CAS:109-99-9] 3000mg/l Dioxan [CAS:123-91-1] 600mg/l Dib 1 * 10 mL
Catalog # CMPLRD04468610.2
Frequently Bought Together